Laboratory Furniture FAQWhat basic information do I need to provide to get a VWR® Furniture quote?Who do I contact for Furniture pricing and availability?Where can I send my orders?Email: vwrfurniture_orders@vwr.com How do I request VWR Furniture Literature?Request a catalog online through our catalog request form. Or, view our online VWR Furniture Literature section. What are the standard heights of casework?VWR offers standing and sitting height casework, 35.75" and 30" respectively for the metal VWR Contour casework. Other heights are available. What are the standard widths of casework?18", 24", 30", 36", 42", 48", 52", and 60" widths. Other custom widths are available. Does VWR provide ADA-compliant casework?Yes, VWR offers ADA-compliant casework, along with ADA-compliant sinks, faucets, fume hoods, and more. What colors for casework are available?Standard colors are #61 Light Neutral and #78 Snow White. Other colors are available upon request. Do the cabinets automatically include worktops?No, worktops are ordered separately as there are many choices of work surfaces, colors, and sizes available to meet your specific needs. What are the standard worktop depths?VWR offers 24" and 31" depths, and 54" and 60" depths for islands and peninsulas. Standard cabinet depth is 22" with a 1" front overhang, which allows a 1" or 8" space at the rear of the cabinet for plumbing and/or electrical. Does VWR provide installation?Yes. Contact your VWR Furniture Specialist for more information. How do I install my own casework?Although we highly recommend that you use one of our highly trained installer; please contact your VWR furniture specialist to request an installation manual. Does VWR offer an energy-conserving, low-flow (high-efficiency) fume hood?VWR offers a variety of fume hoods to meet your requirements. What if I need my lab to be setup quickly?VWR offers a quick-turnaround Rediship Program. Contact your VWR Furniture Specialist for more information. Does VWR offer a turnkey line of laboratory furniture?Yes, please contact your VWR Furniture Specialist for more information. |