Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3e, CD4, CD8a Monoclonal Antibody (PE (Phycoerythrin)-Cy7®, PE (Phycoerythrin), APC (Allophycocyanin)) [clone: 145-2C11, RM4-5, 53-6.7]

Supplier: Tonbo Biosciences
91-0001-U100
10002-342EA 283 USD InStock
10002-342
Anti-CD3e, CD4, CD8a Monoclonal Antibody (PE (Phycoerythrin)-Cy7®, PE (Phycoerythrin), APC (Allophycocyanin)) [clone: 145-2C11, RM4-5, 53-6.7]
Antibodies
The Mouse T Lymphocyte Subset backbone panel consists of 3 anti-mouse antibodies, each conjugated to a different fluorochrome. This panel can serve as a backbone for identifying major T cell subsets, and leaves room for GFP or additional drop-in markers, if desired. A FITC version of this panel is available, where the anti-CD8a antibody is conjugated to FITC.
Hosts: Armenian Hamster, Rat

Type: Primary
Antigen: CD3e, CD4, CD8a
Clonality: Monoclonal
Clone: 145-2C11, RM4-5, 53-6.7
Conjugation: PE (Phycoerythrin)-Cy7®, PE (Phycoerythrin), APC (Allophycocyanin)
Epitope:
Host: Multiple
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty