Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Immunoreagents
GTXRT-003-L2APC
10014-024EA 1894.1 USD InStock
10014-024
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
The ImmunoReagents' Allophycocyanin (APC) is an intensely bright phycobiliprotein isolated from red algae that exhibits far-red fluorescence with high quantum yields. It is excited by laser lines at 594 and 633 nm, with an absorbance maximum at 650 nm and a fluorescence emission peak at 660 nm. APC is a large molecule that is chemically crosslinked to use in fluorescence-based detection, primarily for flow cytometry, microarray assays, ELISAs, and other applications that require high sensitivity but not photostability.

This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty