Would you like to log in again?
Extra Fine Micro Dissecting Scissors 4" Straight Sharp/Sharp, Roboz Surgical
Supplier: Roboz
RS-5882
100189-254EA 121.85 USD InStock
100189-254
Extra Fine Micro Dissecting Scissors 4" Straight Sharp/Sharp, Roboz Surgical
Scissors
Extra Fine Micro Dissecting Scissors; Straight; Sharp Points; 0.4 mm Combined Tip Width; 24 mm Blade Length; 4" Overall Length
These are extra fine dissecting scissors with particularly thin blades and sharp points. The tips are tapered to half the standard size. The combined tip width of these scissors is only 0.4 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty