Would you like to log in again?
Ragnell Scissors 5" Curved Blunt/Blunt, Roboz Surgical
Supplier: Roboz
RS-6015
100189-726EA 185.54 USD InStock
100189-726
Ragnell Scissors 5" Curved Blunt/Blunt, Roboz Surgical
Scissors
Ragnell Scissors; Curved; Slender Blunt Points; 27 mm Blade Length; 5" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty