Would you like to log in again?
Mayo Scissors 9" Curved Sure Cut, Roboz Surgical
Supplier: Roboz
RS-6875SC
100190-330EA 260.31 USD InStock
100190-330
Mayo Scissors 9" Curved Sure Cut, Roboz Surgical
Scissors
Mayo Scissors; Curved; Sure Cut; 63 mm Blade Length ; 9" Overall Length.
Sure Cut scissors make a reliable, clean, smooth cut. There is less force required to cut tissue with these scissors. The bevel-honed edge of Sure Cut scissors acts like a razor edge to cut tissue without crushing it. The serrated edge grips very soft or tough tissue preventing slippage. The firm grip and the superior sharpness guarantee that cutting is always effortless with Sure Cut scissors. WARNING: These scissors feature a razor edge blade which is extremely delicate. They are designed for cutting tissue only. Using these scissors on any hard material such as bone, wire, or fur will res ult in dulling and damaging the blades.
Sure Cut scissors make a reliable, clean, smooth cut
There is less force required to cut tissue with these scissors. The bevel-honed edge of Sure Cut scissors acts like a razor edge to cut tissue without crushing it. The serrated edge grips very soft or tough tissue preventing slippage. The firm grip and the superior sharpness guarantee that cutting is always effortless with Sure Cut scissors. WARNING: These scissors feature a razor edge blade which is extremely delicate. They are designed for cutting tissue only. Using these scissors on any hard material such as bone, wire, or fur will res ult in dulling and damaging the blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty