Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) oxide ≥99.995% (Cu basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Copper monoxide granulatedCopper oxideCupric oxide

29-0590-50G 29-0590-10G
100204-024EA 345.35 USD InStock
100204-024 100209-892
Copper(II) oxide ≥99.995% (Cu basis), PURATREM
Copper (II) oxide


Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1362 °C
Density: 6.3…6.49
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty