Would you like to log in again?
Viscosity Standards, Silicone Oils, Cannon Instruments
Supplier: Cannon Instrument Company
9727-Z42.016 9727-Z30.5 9727-Z38.016 9727-Z10.016 9727-Z30.500 9727-Z26.032 9727-Z30.016 9727-Z30.128 9727-Z50.032 9727-Z42.5 9727-Z10.128 9727-Z26.016 9727-Z46.016 9727-Z10.016 9727-Z42.032 9727-Z30.032 9727-Z18.500 9727-Z14.5 9727-Z50.5 9727-Z22.016 9727-Z26.128 9727-Z18.128 9727-Z34.032 9727-Z14.128 9727-Z34.128 9727-Z38.032 9727-Z34.016 9727-Z30.128 9727-Z26.5 9727-Z14.500 9727-Z22.500 9727-Z42.016 9727-Z46.500 9727-Z34.128 9727-Z10.032 9727-Z22.128 9727-Z18.032 9727-Z22.5 9727-Z46.032 9727-Z38.5 9727-Z42.128 9727-Z26.500 9727-Z22.032 9727-Z14.016 9727-Z42.032 9727-Z26.032 9727-Z38.128 9727-Z42.128 9727-Z38.016 9727-Z34.5 9727-Z18.016 9727-Z10.032 9727-Z38.032 9727-Z18.032 9727-Z46.128 9727-Z50.128 9727-Z50.128 9727-Z14.032 9727-Z30.016 9727-Z22.016 9727-Z14.128 9727-Z18.5 9727-Z34.032 9727-Z38.128 9727-Z34.016 9727-Z50.500 9727-Z46.016 9727-Z22.032 9727-Z10.128 9727-Z46.032 9727-Z14.016 9727-Z30.032 9727-Z18.016 9727-Z26.128 9727-Z10.500 9727-Z18.128 9727-Z26.016 9727-Z34.500 9727-Z50.032 9727-Z14.032 9727-Z10.5 9727-Z50.016 9727-Z38.500 9727-Z46.5 9727-Z50.016 9727-Z22.128 9727-Z46.128 9727-Z42.500
10021-862EA 1158.66 USD InStock
CA10021-870 10021-862 10021-860 10021-902 CA10021-912 CA10021-878 10021-900 CA10021-910 CA10021-876 10021-868 CA10021-874 10021-866 CA10021-872 10021-864 CA10021-908 CA10021-906 CA10021-904 CA10021-902 CA10021-882 10021-872 CA10021-880 10021-870 CA10021-922 10021-914 10021-878 CA10021-888 10021-912 CA10021-920 CA10021-886 10021-876 10021-910 CA10021-884 10021-874 10022-100 10021-908 CA10021-918 10022-102 CA10021-916 10021-906 10021-904 CA10021-914 10022-108 10022-104 10022-106 10021-884 CA10021-892 CA10021-890 10021-882 10021-880 10021-924 CA10021-856 10021-922 CA10021-898 CA10021-854 10021-920 CA10021-896 10021-888 CA10021-852 CA10021-894 CA10021-850 10021-886 CA10022-100 10021-918 10022-098 CA10021-924 10021-916 CA10022-106 CA10022-108 CA10022-102 CA10022-104 10021-850 10021-894 CA10021-860 10021-892 10021-890 CA10021-868 10021-858 CA10021-900 CA10021-866 10021-856 10021-854 10021-898 CA10021-864 10021-896 10021-852 CA10021-862 CA10022-098 CA10021-858
Viscosity Standards, Silicone Oils, Cannon Instruments
Standards Viscometry Standards
CANNON silicone standards are intended for calibration of brookfield type rotational viscometers.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Traceable to NIST.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty