Would you like to log in again?
NZY Broth Dehydrated Media, Teknova
Supplier: Teknova
N1141 N1143 N1142 N1144
100216-976EA 209.23 USD InStock
CA100217-764 CA100219-032 CA100217-906 100216-976
NZY Broth Dehydrated Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For use in the propagation of bacteriophage lambda.
  • Use 20 grams per liter to reconstitute media
Contains 1.0% Select peptone (NZ amine), 0.5% sodium chloride, 0.5% yeast extract.
Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty