Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 5 M, sterile-filtered

Supplier: TEKNOVA INC
Synonyms: HaliteSea saltNaClSaline

S0250 S0250
100219-624EA 123.15 USD InStock
CA100219-624 100219-624
Sodium chloride 5 M, sterile-filtered
Sodium chloride
Suitable for molecular biology, protein chemistry and biochemical applications.


Formula: NaCl
MW: 58.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty