Would you like to log in again?
SOC Broth, Teknova
Supplier: Teknova
S0225
100219-988EA 91.54 USD InStock
100219-988
SOC Broth, Teknova
Media for Microbiology Dehydrated Media for Microbiology
SOC is a nutritionally rich medium used to grow recombinant E.coli
  • Dissolve 32.6 grams per liter of water to reconsitute
  • Autoclave to sterilize
  • Ambient shipping conditions
  • Store at room temperature
Ingredients: 2.0% Tryptone (20 g/L), 0.5% Yeast extract (5.0 g/L), 10 mM Sodium Chloride (NaCl), 2.5 mM Potassium Chloride (KCl), 10 mM Magnesium Chloride (MgCl2), 10 mM Magnesium Sulfate (MgSO4) and 20 mM (0.36%) Glucose (Dextrose) (3.6 g/L).
SOC is the same as SOB, with the addition of Glucose.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty