Would you like to log in again?
VWR® Cleanroom ESD Steel Toe Shoes
Supplier: VWR
VWR®
XY-0816818 XY-0816906 XY-0816809 XY-0816811A XY-0816912 XY-0816803 XY-0816804 XY-0816910 XY-0816902 XY-0816903A XY-0816811 XY-0816907 XY-0816911 XY-0816806 XY-0816905 XY-0816816A XY-0816817 XY-0816907A XY-0816912A XY-0816803A XY-0816812 XY-0816916A XY-0816807A XY-0816813A XY-0816815A XY-0816814 XY-0816916 XY-0816901 XY-0816800 XY-0816913 XY-0816900 XY-0816813 XY-0816810 XY-0816917 XY-0816802 XY-0816801 XY-0816915 XY-0816805 XY-0816812A XY-0816815 XY-0816816 XY-0816903 XY-0816807 XY-0816808 XY-0816904 XY-0816908
75983-206CS 818.63 USD InStock
75983-206 75997-230 10022-210 10022-232 75983-198 10022-234 10022-212 75983-178 10022-230 75983-190 10022-218 75877-376 75983-170 75983-192 75877-370 75983-194 10022-236 10022-214 75877-372 75983-196 75983-174 10022-216 75877-378 75871-634 75983-164 10022-220 75983-186 75983-166 75983-188 10022-222 75983-202 75983-204 10022-228 10022-206 75983-180 10022-208 75879-252 75983-182 10022-224 75983-160 75877-380 10022-226 10022-204 75983-162 75983-184 75877-382
VWR® Cleanroom ESD Steel Toe Shoes
Shoes and Boots
VWR® now offers new professional safety shoes made from synthetic leather upper for comfort, durability, and abrasion resistance. In addition, these shoes feature electrical hazard protection to enhance safety in the workplace.
  • Electrostatic discharge protection
  • Synthetic leather uppers
  • Non-skid polyurethane sole
  • Protective steel toe cap
High quality construction provides a comfortable cushioning effect and helps keep feet dry, by allowing moisture to escape at a rapid rate. The safety shoe is completely lined with a durable and breathable lining to keep feet cooler. The injected polyurethane sole is resistant to acids and oils, which is important for many production environments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty