Would you like to log in again?
Creatinine Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
C7539-150 HC939-1340 OC939-739 HC939-756 C7539-1250 C7539-625
10023-482EA 699.9 USD InStock
10023-482 10022-998 10022-624 10022-626 10022-950 10022-622
Creatinine Reagent Set, Pointe Scientific
Assays Cellular Assays
Creatinine reagent is used for the quantitative determination of creatinine in serum.
  • Quantitative determination of creatinine in serum
  • In vitro diagnostic use only
Creatinine reagent is used for the quantitative determination of creatinine in serum
Hitachi Creatinine reagent (739 total mL)is used on the Hitachi 917 analyzer only and is for in vitro diagnostic use only. Creatinine reacts with picric acid in alkaline conditions to form a color complex that absorbs at 510 nm. The rate of formation of color is proportional to the creatinine in the sample. There are (9) 67 mL bottles of R1 Reagent and (9) 17 mL bottles of R2 reagent in the 739 total mL kit.
The 1340 total mL kit is used on Hitachi 717, 911, 912, and 914 analyzers. For the quantitative determination of creatinine in serum on Hitachi analyzers. For in vitro diagnostic use only. Creatinine reacts with picric acid in alkaline conditions to form a color complex that absorbs at 510 nm. The rate of formation of color is proportional to the creatinine in the sample. There are (11) 100 mL bottles of R1 Reagent and (3) 80 mL bottles of R2 Reagent in this kit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty