Would you like to log in again?
Fluoromount-G® Slide Mounting Medium, Southern Biotech
Supplier: Southern Biotechnology
Fluoromount™
0100-01
100241-874EA 58.85 USD InStock
100241-874
Fluoromount-G® Slide Mounting Medium, Southern Biotech
Stains Microscopy and staining reagents
Fluoromount-G® is suitable for mounting frozen and paraffin sections, whole mounts, and cells.
  • For mounting frozen and paraffin sections, whole mounts, and cells
  • Water soluble
  • Semi-permanent seal
Fluoromount-G® is a water soluble compound recommended for slides mounted after a staining procedure having an aqueous final step. Because Fluoromount-G® is water soluble, the coverslip may be removed by submerging the slide in a PBS solution until the coverslip is loosened. This mounting medium also provides a semi-permanent seal for storage of slide preparations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty