Would you like to log in again?
Whatman™ Puradisc Syringe Filters, CA, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Puradisc™ Puradisc™
10462601 10462701 6771-1304 10462100 10462701 10462701 10462205 10462700 10462600 10462600 10462240 10462610 10462260 10462100 10462200 6771-1304 10462601 10462205 10462700 10462243 10462610 10462710 10462261 10462243 10462200 10462263 10462263 10462241 10462710 10462206 10462206 10462260 10462261 10462240 10462241
10035-806PK 1287.25 USD InStock
10035-806 CA11013-476 89233-774 CA14224-474 10035-804 89233-776 14224-468 CA11013-154 89233-772 CA11013-150 CA11008-550 10035-092 10035-090 28151-261 CA11012-946 10035-096 10035-094 10035-112 CA11013-926 CA10033-472 CA28151-261 CA89233-776 CA11013-126 CA89233-774 CA89233-772 CA11013-122 14224-474 CA11012-962 11008-550 CA11012-944 CA14224-468 CA10462701 10035-088 10035-104 10035-504
Whatman™ Puradisc Syringe Filters, CA, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • From the Puradisc FP 30 range
  • Eight different material options
  • Choice of 4, 13, 25 and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample types and applications, including capillary electrophoresis, biological preparations, protein filtration, and UV spectroscopy sample preparation. Different Puradisc membrane media support diverse chemical solvents and compounds.
Puradisc is available in the following membrane materials: regenerated cellulose (RC), cellulose nitrate, nylon, polytetrafluoroethylene (PTFE), polyethersulfone (PES), polypropylene, polyvinylidene difluoride (PVDF), and cellulose acetate.
Cytiva's Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of applications. Cytiva also offers high-performance GD/X syringe filters with a multi-layer glass fiber prefilter for filtering larger volumes of harder-to-filter samples.
Cellulose acetate syringe filters exhibit very low protein binding, so they are suitable for protein recovery applications. They are hydrophilic, making them functional with aqueous and alcoholic media although they have limited solvent resistance.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty