Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRRC32 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Plato-1]

Supplier: Enzo Life Sciences
804-867FI-0100
10049-882EA 590 USD InStock
10049-882
Anti-LRRC32 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Plato-1]
Antibodies
LRRC32 (leucine rich repeat containing 32; also known as GARP or Garpin; Glycoprotein A repetitions predominant) is a glycoprotein expressed on the cell surface of megakaryocytes, platelets and activated regulatory T (Treg) cells.

Type: Primary
Antigen: LRRC32
Clonality: Monoclonal
Clone: Plato-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty