Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP20 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-796-D
10050-184EA 280 USD InStock
10050-184
Anti-HSP20 Rabbit Polyclonal Antibody
Antibodies
Hsp20 is a member of the family of small heat shock proteins (sHsps), with homology to Hsp25, Hsp27, and alphaB-crystallin. Highest expression of Hsp20 is found in skeletal, smooth, and cardiac muscle. Under heat shock, Hsp20 translocates from the cytosol to the nucleus, although the amount of Hsp20 in the cell does not increase like other Hsps. Hsp20 oligomerization and interaction with polymeric actin filaments is regulated via phosphorylation by PKA and PKG.

Type: Primary
Antigen: HSP20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Fish, Dog, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty