Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutaraldehyde 10% in aqueous solution, EM Grade

Supplier: Electron Microscopy Sciences
Danger
Synonyms: 1,5-PentanedialGlutardialdehydeGlutaral

16100 16120 16110 16122 16121
100504-786BX 65.66 USD InStock
100504-786 100503-976 100503-974 100503-972 100503-970
Glutaraldehyde 10% in aqueous solution, EM Grade
Glutaraldehyde
After glutaraldehyde has been purified to EM grade, it has a high risk of polymerizing if it is not handled properly. Electron Microscopy Sciences takes special precautions to assure a polymer-free, pure glutaraldehyde. Every lot of our EM grade aqueous glutaraldehyde is assayed prior to shipping assuring you consistent purity and the highest possible quality. If stored properly, our Glutaraldehyde has a very long shelf life.

  • Specific gravity 20/20 °C: 1.131
  • Distillation purified

EM grade aqueous glutaraldehyde, stored in volumes greater than 100 ml, with repeated defrosting and recapping, will polymerize in time, producing an absorption peak of 235 nm rather than the 280 nm of pure EM grade. The presence of polymerized glutaraldehyde greatly reduces its efficiency as a cross-linking agent, preventing proper fixation. For this reason, we provide 10 ml ampoules and 100 ml bottle quantities for larger applications.

Packaging: We package our aqueous glutaraldehyde in 10 ml, single dose ampoules, sealed under dry nitrogen, and kept refrigerated until it is shipped. We offer our glutaraldehyde in a wide range of percentages to simplify mixing procedures and avoiding waste. 1 box contains 10x10 ml ampoules.


Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00007025
CAS Number: 111-30-8
UN: 3082
ADR: 9,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty