SERVICES
Services: VWR CATALYST™ offers recertification, calibration, and repair services for all makes and models of thermometers. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for quote today. Our thermometer services include: Complete visual inspection for any physical damage. If possible, the damage will be repaired. Recertification from 1 Point to 15 Points. Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures. Each temperature is recorded to 1/10th of a scale division. A calibration report is generated, detailing pertinent information including the correction factors for each test point. All calibration information is entered into a data storage system. All certification procedures are performed in accordance with NIST and ASTM guidelines. |