Would you like to log in again?
Extraction Thimble Filters, Advantec MFS
Supplier: Advantec
N08425X29X60MM N08425X29X80MM N088RH22X25X90MM N088R19X23X90MM N08428X32X100MM N08443X123MM N08430X34X80MM N08421X25X90MM N088R30X34X100MM N089S22X25X90MM N088R30X34X80MM N08443X47X123MM N08922X25X90MM N086R19X23X90MM N08433X37X100MM N08430X34X100MM N08419X22X90MM N086R30X34X100MM N08425X29X100MM N08422X25X80MM N086R21X25X90MM N08422X25X65MM N08433X37X80MM N086R30X34X80MM N08425X29X90MM N086R43X47X123MM N086R25X30X100MM N088R25X29X100MM
101102-536PK 101.94 USD InStock
101102-536 101102-614 101102-318 101102-534 101102-452 101102-298 101102-454 101102-392 101102-394 101102-450 102830-240 102830-242 101102-466 101102-620 101102-500 101102-588 101102-622 101102-468 101102-462 101102-386 101102-464 101102-388 101102-586 101102-460 101102-390 102830-238 101103-624 CA101102-450
Extraction Thimble Filters, Advantec MFS
Extractors Extraction Thimbles
High purity, seamless thimble filters offered in 5 materials
Cellulose Fiber–Made from alpha cotton cellulose, these thimbles are suitable for Soxlet extractions of organic components and dust sampling. Lipid content: <0.1% wt.
Glass Fiber–Made of Borosilicate glass, these thimbles can withstand temperatures up to 500°C making them ideal for high temp. extractions or dust monitoring. They are acid resistant (except hydrofluoric). Thimbles have been acid washed for minimum trace metal content.
Silica Fiber(Quartz)–Made of pure quartz fibers, these thimbles can be used in temperatures up to 1000°C. Tapered for ease in loading into stack samplers. Heat-treated, Item 101102-460 is pretreated at 900°C for 4hrs to stabilize weight.
PTFE Fiber and PTFE/Silica Fiber: PTFE is inherently hydrophobic and non-absorptive with respect to acid gases. Negligible to no trace metal contamination. Durable and temperature- resistant.
Applications include analysis of fats, oils, grease, pesticides, pollutants, organics and addititives in plastics and rubbers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty