Would you like to log in again?
Sani-Plex 128 Detergent, Quiplabs
Supplier: Quip Laboratories
Sani-Plex 128
S128M-CASE
10121-284CS 873 USD InStock
10121-284
Sani-Plex 128 Detergent, Quiplabs
Cleaning Agents Detergents and Surface Cleansers
Sani-Plex 128 is a multipurpose, neutral pH, germicidal detergent that disinfects and cleans in one labor-saving step.
It is effective in hard water in the presence of a moderate amount of soil (5% organic serum) as calculated by the AOAC Use-dilution Test.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty