Would you like to log in again?
EXP® Trap Cartridge 5.0 µm EXP
Supplier: Optimize Technologies
EXP®
15-03964-HP 15-03978-HU 15-03973-HV 15-03969-HR 15-03964-HR 15-03973-HQ 15-03964-HU 15-03983-HS 15-03978-HV 15-03983-HQ 15-03964-HT 15-03973-HU 15-03969-HS 15-03973-HP 15-03978-HP 15-03983-HR 15-03964-HS 15-03978-HT 15-03964-HQ 15-03973-HT 15-03969-HP 15-03978-HQ 15-03969-HT 15-03978-HS 15-03973-HS 15-03973-HR 15-03978-HR 15-03983-HU 15-03983-HT 15-03969-HQ 15-03969-HU 15-03983-HV 15-03964-HV 15-03983-HP 15-03969-HV
10123-300PK 1118.45 USD InStock
10123-300 10123-288 10123-302 10123-304 10123-306 10122-816 10122-814 10122-818 10123-286 10122-820 10122-824 10122-802 10122-800 10122-822 10123-310 10123-312 10123-292 10122-828 10122-806 10123-294 10122-826 10122-804 10123-296 10123-298 10122-808 10122-798 10122-796 10122-812 10122-756 10123-290 10122-810 10122-790 10123-308 10122-794 10122-792
EXP® Trap Cartridge 5.0 µm EXP
Chromatography Columns
The patented hand-Tight EXP® trap column is rated for use at 20000+ psi (1400+ bar). This unique design connects directly to any injection valve (with 10-32 threads) or in-line with ¹/₁₆" stainless tubing for unparalleled convenience and efficiency.
  • Rated to 20000+ psi (1400+ bar)
  • Bed volumes from 1.6 to 600 μl
  • Convenient means of handling a variety of sample clean-up and enrichment processes
  • More efficient than off-line trapping techniques
  • Hand-tight trap replacement - no tools required
The EXP® cartridge system enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn® architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet.
Ordering information: Custom packing available.
Delivery information: Includes 3 uni-directional cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty