Would you like to log in again?
Sartorius Biohit Family Pipette Calibration Depot Services, Sartorius
Supplier: Sartorius
84LOP10 84LOP10 84KMP01 84KOP05 84LOP02 84LMP01 84JEP12 84LMP04 84JMP01 84KOP05 84JMP02 84KOP12 84KMP02 84JEP05 84JEP12 84KOP04 84JMP05 84LMP05 84KMP02 84JOP05 84LOP04 84LOP02 84JEP02 84JOP12 84JOP05 84JMP10 84KOP01 84JOP12 84LOP12 84KMP01 84JOP10 84LMP02 84JOP10 84JOP02 84KMP12 84LOP01 84LMP01 84LOP04 84JMP12 84KOP10 84KOP02 84LMP10 84KMP05 84LMP10 84KMP10 84LMP12 84LOP05 84JEP01 84JEP01 84JOP01 84JEP10 84JMP04 84LMP02 84KOP10 84JEP02 84JOP01 84KMP10 84JMP05 84LOP01 84KMP05 84KOP01 84JMP02 84KOP12 84JEP10 84KMP04 84LOP05 84JEP04 84JEP04 84JMP04 84JMP10 84KOP04 84LOP12 84JMP12 84KMP12 84LMP05 84JOP02 84KOP02 84JOP04 84KMP04 84JOP04 84JEP05 84LMP04 84LMP12 84JMP01
75995-120EA 164.4 USD InStock
75995-120 75995-122 CA10129-118 CA10129-116 CA10129-238 10129-268 CA10129-236 CA10129-234 CA10129-278 10129-276 CA10129-242 10129-232 CA10129-122 10129-234 CA10129-240 CA10129-120 10129-272 10129-194 10129-274 10129-230 10129-196 10129-190 10129-270 75995-118 10129-192 CA10129-268 10129-136 CA10129-266 10129-138 10129-188 CA10129-276 CA10129-232 CA10129-196 10129-266 CA10129-230 CA10129-274 10129-184 CA10129-194 10129-140 CA10129-272 10129-186 CA10129-192 10129-142 CA10129-270 CA10129-190 10129-180 10129-182 CA10129-138 CA10129-136 10129-126 10129-246 10129-128 CA10129-178 CA10129-134 10129-248 CA10129-188 10129-132 CA10129-142 10129-134 CA10129-186 CA10129-140 CA10129-184 10129-130 CA10129-182 CA10129-180 10129-118 CA10129-248 CA10129-128 10129-236 CA10129-246 CA10129-126 10129-238 CA10129-244 10129-116 CA10129-124 10129-122 CA10129-132 CA10129-176 10129-242 10129-124 CA10129-130 10129-244 10129-120 10129-240
Sartorius Biohit Family Pipette Calibration Depot Services, Sartorius
Services
Sartorius Liquid Handling mail-in pipette calibration services are designed to provide high-quality pipette calibration with simplified options.
  • Computer-Generated Calibration Certificates
  • Evaluation Against Manufacturer’s Known Tolerances for Systematic and Random Error
  • Pipettes Receive a Calibration Sticker with the Service Date and Next Recommended Calibration Date
  • Secure Transit Cartons for Each Shipment
  • Simplified Service and Decontamination Forms
Pipet services feature computer-generated calibration certificates, an evaluation against manufacturer’s known tolerances for systematic and random error, a calibration sticker with the service date and next recommended calibration date for pipets, secure transit cartons for each shipment, and simplified service and decontamination forms
Users can choose from three types of services: Calibration with Cleaning and Lubrication, Calibration with Critical-Part Replacement (for Biohit/Sartorius pipets only), and Preventative Maintenance.
All models receive a visual leak test, and all multi-channel units receive a linearity check.
Certifications: All calibration equipment and certificates are ISO compliant and traceable through NIST. ISO 17025 Accredited Calibration Laboratory.
Accessories information: Additional “As Found” performance checks are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty