Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Exhaust Filter Kit
Supplier: Revvity Health Sciences
N8145005
10136-754EA 204.75 USD InStock
10136-754
Exhaust Filter Kit
Filters Venting Filters
he exhaust filter kit is designed to be used with Leybold SOGEVAC SV40BI vacuum pumps.
Filter should be inspected and replaced perodically as a part of regular maintenance of the vacuum pump.
Delivery information: Complete exhaust filter cartridge kit comes with required O-ring and fits Leybold SOGEVAC SV40BI vacuum pumps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty