Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% in aqueous solution normal saline solution, sterile

Supplier: TEKNOVA INC
Synonyms: HaliteSea saltNaClSaline

S5812 S5815 S5825 S5819 S5820 S5822 S5825 S5819 S5815 S5825 S5820 S5821 S5827 S5812
101320-574PK 155.93 USD InStock
101320-574 101399-918 101399-916 CA100216-354 101399-914 76423-606 CA101320-570 76423-602 76423-604 77442-984 CA101320-568 100216-354 CA101320-574 CA101320-572
Sodium chloride 0.9% in aqueous solution normal saline solution, sterile
Sodium chloride
Dionized Water used as a solvent or diluent for antibiotics and biologicals.


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty