Would you like to log in again?
Dulbecco's Phosphate-Buffered Saline (DPBS)
Supplier: PromoCell
C-40230 C-40232
10172-542EA 32.08 USD InStock
10172-542 10172-540
Dulbecco's Phosphate-Buffered Saline (DPBS)
Buffers Cell Culture Buffers
These Dulbecco's PBS solution are used in tissue culture, cell isolation procedures and molecular biology.
  • Ready-to-use buffers
  • Storage at 4 to 8 °C
Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty