Would you like to log in again?
Museux Vulsellum Forceps, OR Grade, Sklar
Supplier: Sklar
91-3006
10192-282EA 339.56 USD InStock
10192-282
Museux Vulsellum Forceps, OR Grade, Sklar
Forceps
Museux Vulsellum Forceps are ratcheted, finger ring forceps used for grasping the uterus during OB/GYN procedures.
The shanks curve slightly inward after the ratchet mechanism providing a long, narrow reach.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty