Would you like to log in again?
Sklarlite™ Baumgartner Needle Holder, OR Grade, Sklar®
Supplier: Sklar
23-2132
10195-328EA 259.79 USD InStock
10195-328
Sklarlite™ Baumgartner Needle Holder, OR Grade, Sklar®
Forceps
Sklarlite™ Baumgartner Needle Holders feature heavy serrations which make this instrument capable of firmly holding larger needles and dense suture material
They are commonly used in procedures on the trunk and limbs. Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue. This product is straight with serrated tips and a length of 5-1/4 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty