Would you like to log in again?
VWR® P Portable Balances
Supplier: VWR
VWR®
VWR-6000P VWR-123P VWR-1500P VWR-3000P VWR-500P
10205-006EA 660.05 USD InStock
10205-006 10205-008 10205-010 10205-002 10205-004
VWR® P Portable Balances
Balances and Scales
The VWR® P-Series Balances are ideal for educational, light industrial, and general laboratory applications that require accuracy and portability.
  • Portable design
  • Stainless steel weigh pan
  • Rugged construction
  • External calibration
  • Five-year warranty
Portable balances offer simple operation and multiple weigh modes. All units feature a stainless steel weigh pan. They are rugged and built to last.
Delivery information: VWR-123P (10205-002) includes 100 g calibration weight.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty