Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Single and Multi Wafer Containers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64918-4 64917-2 64918-2 64917-4 64918-3 64917-46 64917-25 64917-1 64917-3 64917-6
102095-792PK 82.08 USD InStock
102095-792 102095-794 102095-804 102095-790 102095-800 102095-802 102095-786 102095-796 102095-788 102095-798
Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Handling Supplies Wafer Carriers
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
  • Conical base to support the wafers on their edges
  • Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
Delivery information: The set includes the container, the cover and the spring.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty