Would you like to log in again?
Phosphate buffered saline, tablets, Novus Biological
Supplier: Novus Biologicals
NB900-62086
102136-946EA 309.97 USD InStock
102136-946
Phosphate buffered saline, tablets, Novus Biological
Buffers Electrophoresis Buffers Washing Buffers
Phosphate Buffered Saline Tablets pH 7.4
Dilutions: Immunohistochemistry 1:10-1:500
Application Notes: Dissolve one tablet in 100 ml deionized water. The resulting solution contains 137 mM Sodium Chloride, 2.7mM Potassium Chloride, 10mM Phosphate Buffer, at a pH of 7.4 +/- 0.1
Ordering information: This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty