Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DUSP23 Mouse Monoclonal Antibody [clone: 3C10]

Supplier: Novus Biologicals
NBP2-01507
102229-750EA 663.82 USD InStock
102229-750
Anti-DUSP23 Mouse Monoclonal Antibody [clone: 3C10]
Antibodies
The DUSP23 Antibody (3C10) from Novus Biologicals is a mouse monoclonal antibody to DUSP23. This antibody reacts with human. The DUSP23 Antibody (3C10) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DUSP23
Clonality: Monoclonal
Clone: 3C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty