Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alera® Traditional Series Guest Arm Chair

Supplier: Alera
Traditional
ALETD4336 ALETDC4336 ALETDC4336
102830-232EA 750.26 USD InStock
102830-232 500005-217 102830-230
Alera® Traditional Series Guest Arm Chair
Chairs
Handsomely appointed and attractively priced for the fiscally discerning executive.

  • Traditional "bankers" styling with brass nailhead trim
  • Deep cushioned vinyl upholstery
  • Genuine hardwood frame with a rich mahogany finish
  • Vinyl padded hardwood arms for comfort and support
  • Straight wood legs

Arm height from seat 6.25"
Distance between arms 22.5"

Certifications: GREENGUARD Indoor Air Quality Certified®, Meets or exceeds ANSI/BIFMA standards

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty