Would you like to log in again?
ELISABright, Advansta
Supplier: Advansta
K-16025-D10 K-16025-D25
103254-876EA 317.31 USD InStock
103254-876 103254-920
ELISABright, Advansta
Stains Microscopy and staining reagents
Superior chemiluminescent substrate for ELISA applications.
  • Highest signal-to-noise ratio for superior performance
  • Broad dynamic range for enhanced detection and precision
  • Immediate light generation for fast detection and results
  • Cost effective with reduced reagent consumption
ELISABright is an enhanced and highly sensitive chemiluminescent HRP substrate optimized for chemiluminescent ELISA applications. This luminol-based substrate demonstrates a wide linear dynamic range (Figure 1) and a superior signal to noise ratio (Figure 2) for excellent quantitative ability. ELISABright allows for enhanced detection of low abundance proteins and reduced consumption of antibodies and other reagents.
Ordering information: Includes ELISABright luminol and ELISABright peroxide reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty