Would you like to log in again?
Mag4C Conservation Buffer, OZ Biosciences
Supplier: OZ Biosciences
LVB1000 ADB1000
103258-266EA 124.62 USD InStock
103258-266 103258-264
Mag4C Conservation Buffer, OZ Biosciences
Buffers Biological Buffers
Mag4C Conservation Buffer can be added to viral stocks for long-term storage
  • Preserves virus infectivity during storage
  • Minimizes the loss of viral titers during storage and freeze/thaw cycles
  • Can be used to conserve viruses bound to Mag4C beads
It allows maintaining virus infectivity even after multiple freeze/thawing cycles. Two different buffers are available depending on the virus: Ad-Conservation Buffer for adenoviruses & Lv-Conservation Buffer for lentiviruses and retroviruses.
Delivery information: Shipped at room temperature
Packaging: Store at +4°C
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty