Would you like to log in again?
Bradford Protein Assay Kit (B-Pak), OZ Biosciences
Supplier: OZ Biosciences
BA00050 BA00070 BA00100
103258-314EA 147.69 USD InStock
103258-314 103258-318 103258-316
Bradford Protein Assay Kit (B-Pak), OZ Biosciences
Assays Protein Assays
Bradford Protein Assay Kit (B-Pak) is a straightforward and rapid kit for determining the concentration of proteins in solution.This ready-to-use kit is provided with 1X reagent & 2 sets of BSA standards which means that no dilution, filtration or calculation are required
  • Accurate determination of protein concentration
  • Ready to use prediluted standard protein
  • Cheaper than competitors
  • Convenient packaging
  • Fast and easy-to-use Bradford protein assay
Simply process your assay in a few minutes. B-Pak is provided with adapted protocols for all experimental conditions: micro and macroassays.
Delivery information: Shipped at room temperature
Packaging: Store at +4°C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty