Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FBN1 Mouse Monoclonal Antibody [clone: 3H6]

Supplier: Novus Biologicals
H00002200-M01
103326-242EA 798.46 USD InStock
103326-242
Anti-FBN1 Mouse Monoclonal Antibody [clone: 3H6]
Antibodies
The Fibrillin 1 Antibody (3H6) from Novus Biologicals is a mouse monoclonal antibody to Fibrillin 1. This antibody reacts with human. The Fibrillin 1 Antibody (3H6) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: FBN1
Clonality: Monoclonal
Clone: 3H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty