Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FBN1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46120
103408-488EA 564.74 USD InStock
103408-488
Anti-FBN1 Goat Polyclonal Antibody
Antibodies
The Fibrillin 1 Antibody from Novus Biologicals is a goat polyclonal antibody to Fibrillin 1. This antibody reacts with human. The Fibrillin 1 Antibody has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence, Peptide ELISA.

Type: Primary
Antigen: FBN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal region
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty