Would you like to log in again?
Kwazar® Dual-Action Premium GHS Spray Bottle, HCL Labels
Supplier: HCL Labels
GHSKWBOT0009 GHSKWBOT0020 GHSKWBOT0024 GHSKWBOT0021 GHSKWBOT0007 GHSKWBOT0033 GHSKWBOT0005 GHSKWBOT0034 GHSKWBOT0010 GHSKWBOT0014 GHSKWBOT0027 GHSKWBOT0030 GHSKWBOT0031 GHSKWBOT0029 GHSKWBOT0008 GHSKWBOT0001 GHSKWBOT0016 GHSKWBOT0025 GHSKWBOT0032 GHSKWBOT0018 GHSKWBOT0012 GHSKWBOT0002 GHSKWBOT0015 GHSKWBOT0026 GHSKWBOT0035 GHSKWBOT0006 GHSKWBOT0037 GHSKWBOT0028 GHSKWBOT0036 GHSKWBOT0017 GHSKWBOT0013 GHSKWBOT0011
103538-328EA 18.54 USD InStock
103538-328 103538-326 103538-266 103538-288 103538-320 103538-286 103538-264 103538-324 103538-268 103538-322 103538-280 103538-284 103538-262 103538-282 103538-260 103538-332 103538-276 103538-254 103538-330 103538-274 103538-252 103538-296 103538-258 103538-334 103538-278 103538-256 103538-290 103538-250 103538-272 103538-294 103538-270 103538-292
Kwazar® Dual-Action Premium GHS Spray Bottle, HCL Labels
Bottles Spray Bottles
Pre-labeled Kwazar® GHS Spray Bottles meet all GHS requirements and are made from chemical-resistant, laboratory grade HDPE material.
  • Ergonomic dual action trigger
  • Viton reinforced sprayer
  • Labels are made of a durable 3M adhesive vinyl, laminated for chemical and solvent resistance
  • Bottles are supplied empty, and ready to be filled
The premium bottles are sure to meet all laboratory spray bottle needs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty