Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Monoclonal Antibody

Supplier: Abnova
MAB8175
10720-398EA 527.69 USD InStock
10720-398
Anti-FTO Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of Fto.

Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: Fto
Clonality: Unconjugated
Clone: Monoclonal
Conjugation:
Epitope:
Host: IgG
Isotype:
Reactivity: Bovine, Dog, Horse, Human, Mouse, Non-Human Primates, Rat, Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty