Would you like to log in again?
ASTM Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Supplier: Thermco
ACC109F ACC072C ACC120C ACC113C ACC117C ACC127C ACC132C ACC110C ACC097F ACC129C ACC128C ACC114C ACC130C ACC078F ACC098F ACC129F ACC108F ACC123C ACC077F ACC068C ACC125C ACC081F ACC121C ACC070F ACC072F ACC128F ACC079F ACC068F ACC069F ACC136F ACC111C ACC130F ACC126C ACC097C ACC124C ACC098C ACC110F ACC116C ACC113F ACC080F ACC118C ACC112C ACC126F ACC070C ACC136C ACC069C ACC118F
10747-288EA 61.99 USD InStock
10747-288 10747-266 10747-244 10747-344 10747-322 10747-366 10747-368 10747-346 10747-324 10747-226 10747-326 10747-228 10747-370 10747-350 10747-372 10747-230 10747-374 10747-352 10747-232 10747-310 10747-354 10747-332 10747-234 10747-312 10747-334 10747-356 10747-258 10747-236 10747-314 10747-336 10747-358 10747-316 10747-338 10747-318 10747-260 10747-282 10747-360 10747-284 10747-262 10747-362 10747-384 10747-242 10747-286 10747-264 10747-364 10747-386 10747-320
ASTM Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil and petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Ordering information: Accessories not included. Certificate of calibration not included. ASTM thermometers with a NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory are also available; contact your Avantor representative for more information.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers recertification, calibration, and repair services for all makes and models of thermometers. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for quote today.
Our thermometer services include: Complete visual inspection for any physical damage. If possible, the damage will be repaired. Recertification from 1 Point to 15 Points. Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures. Each temperature is recorded to 1/10th of a scale division. A calibration report is generated, detailing pertinent information including the correction factors for each test point. All calibration information is entered into a data storage system. All certification procedures are performed in accordance with NIST and ASTM guidelines.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty