Ultra stack hanging bench racks are used to hang Quantum ultra bins in a range of configurations that can meet personal storage requirements. These bench racks have a 16 gauge, cold rolled steel construction.
- Weight capacity up to 2000 on large louvered panel systems
- Gray baked-enamel finish ensures years of heavy duty use
- Different racks will accept different sizes of bin
Ultra Hanging Systems Quantum Ultra bins will hang on bench racks in a wide range of configurations designed to meet all application needs. Table top Bench Racks are formed from heavy duty 16 gauge cold rolled steel and have a Gray baked-enamel finish which ensures years of heavy duty use.
Ordering information: Bins can be acquired in a variety of sizes and colors (sold separately).