Would you like to log in again?
Hypochlorite WFI Sterile Solutions, STERIS®
Supplier: Steris
1S0696WR 1S0677WR 1S0696WR 1S0896WR 1S0808WR 1S0677WR 1S0877WR 1S0608WR 1S0608WR 1S0877WR 1S0808WR 1S0896WR
10755-284CS 649.16 USD InStock
10755-284 CA75780-040 10755-286 10755-288 CA75780-056 CA75780-038 CA75780-058 CA75780-036 10755-290 10755-282 CA75780-028 10755-292
Hypochlorite WFI Sterile Solutions, STERIS®
Cleaning Agents Detergents and Surface Cleansers
STERIS' 0.525% or 5.25% Hypochlorite WFI sterile solution removes residue from hard surfaces even from the most critical surfaces. Oxidizing cleaners can be extremely effective with tough soils.
  • Quality - Extremely low level of endotoxins from WFI produced by a Finn-Aqua® Multiple Effect Water Still
  • Effective - High quality sodium hypochlorite solution oxidizes difficult to remove soils
  • Convenience - Double bagged for easy introduction into your aseptic areas
  • Safety - 0.2 micron filtered, aseptically filled and packaged minimizes potential for particles being introduced during use
  • Assurance - Each lot documented by a Certificate of Analysis including sterility results
STERIS' 0.525% or 5.25% Hypochlorite WFI sterile solution is made with United States Pharmacopeia (USP) quality Water For Injection and Sodium Hypochlorite. It is designed to remove residues from surfaces. Sterile filtered and filled in an ISO Class 5 aseptic environment, each package is double bagged for easy entry into your controlled environments. This product has an extensive documentation package to meet your validation objectives and is supported by the STERIS Technical Services team for individualized application and validation assistance.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty