The Zeitfuchs Cross-Arm Viscometer measures kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445 and ISO 3104
Viscometer is calibrated. The unity factor series (ZCAXC) have a constant within ±0.2% of the listed value. Order with permanently attached round holder (-RO), rectangular holder (-RE) or order holder separately. Minimum sample volume 1 to 3 mL. Minimum bath depth of 9".