Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Unifine® Pentips®, Owen Mumford
Supplier: Owen Mumford
Unifine® Pentips®
AN3530 AN3840 AN3590 AN3529 AN3550
10804-703CS 121.76 USD InStock
10804-703 10804-705 10804-707 10804-709 10804-711
Unifine® Pentips®, Owen Mumford
Blood Lancets
Unifine® Pentips® are available in five needle lengths (4mm, 5mm, 6mm, 8mm, and 12mm), and are compatible with all diabetes medication pens.
Certifications: CE compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty