Would you like to log in again?
VIALS 300UL CLR VIALS&PTFE/SIL/PTFE 9MMD
Supplier: MicroSolv Technology Corp
Vial^ Easy Purchase Pack Vial ScrewTop and Blue Screw Caps with PTFE/Silicone/PTFE. Dimension: 12x32mm, Fused Inserts. Material: Borosilicate, Color: Clear, Deactivated. 300ul
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIALS 300UL CLR VIALS&PTFE/SIL/PTFE 9MMD 10804-934 9532S-2DCP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty