Would you like to log in again?
I-300 PRO VACUBOX WOULFBOTL
Supplier: Buchi
Interface^ Touch Interface I-300 Pro controls all distillation parameters of a Rotavapor R-300 system, Modes: manual, timer, continuous, AutoDest, drying, methods, Charting, recording, mobile app available, Features: Interface: I-300 Pro Compatibility Pack: R-300, V-300
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
I-300 PRO VACUBOX WOULFBOTL 10806-454 11I300P20 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty