Would you like to log in again?
SUMO-QAPTURE-T® Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
UW1000A-0001
10810-052EA 511 USD InStock
10810-052
SUMO-QAPTURE-T® Kit, Enzo Life Sciences
Assays Protein Assays
The only commercially available kit for isolation and enrichment of SUMOylated proteins.
  • Fast, simple, and reliable assay
  • High specificity, high through-put capacity
  • Amenable to analysis via Western blotting or proteomic methods
For the isolation and enrichment of SUMOylated proteins.
Suggested Applications include: Capture and isolation of SUMO-protein conjugates from specific cell/tissue lysates of interest with subsequent detection and analysis by Western blotting. Identification of SUMO-modified protein substrates by proteomic analysis methods following release of free SUMOylated proteins in denatured form. Selective purification/pull down of SUMOylated proteins from in vitro SUMOylation assays.
The SUMO-QAPTURE-T® kit is an efficient tool for the selective isolation of SUMOylated proteins. The kit facilitates the affinity purification of SUMOylated proteins from cell extracts and tissue lysates using a high-binding SIM-containing affinity matrix. Captured proteins are eluted under denaturing conditions followed by analysis by Western blotting, using the SUMO antibodies provided or antibodies to specific proteins of interest, or potential substrate identification by proteomic methods. SUMO-conjugate-containing samples are prepared in native form in the presence of protease inhibitors to prevent loss through the action of deSUMOylating enzymes. Optimization of binding permits complete isolation of the full range of SUMO-protein conjugates from a specific lysate.
Ordering information: Contains SUMO-QAPTURE-T® matrix, Recombinant SUMO-1, 2 and 3 protein controls, SUMO antibody solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty