Would you like to log in again?
Microchem® by AlphaTec™ 68-4000 Chemical Protection Coveralls with Hood and Ultrasonically Welded and Taped Seams, Ansell
Supplier: Ansell
GR40-T-92-111-05 GR40-T-92-111-08 GR40-T-92-122-09 GR40-T-92-122-05 GR40-T-92-122-08 GR40T9211107 GR40-T-92-122-06 GR40-T-92-111-04 GR40-T-92-111-09 GR40-T-92-111-06 GR40-T-92-122-04 GR40-T-92-111-03 GR40-T-92-111-02 GR40-T-92-122-02 GR40-T-92-122-03 GR40-T-92-122-07
10836-246CS 457.14 USD InStock
10836-246 10836-520 10836-234 10836-442 10836-244 10836-514 10836-238 10836-524 10836-248 10836-236 10836-522 10836-518 10836-516 10836-440 10836-242 10836-240
Microchem® by AlphaTec™ 68-4000 Chemical Protection Coveralls with Hood and Ultrasonically Welded and Taped Seams, Ansell
Disposable Garments
These coveralls are designed to provide an exceptional barrier against organic and inorganic chemicals as well as biological agents.
  • Permeation tested against over 190 chemicals, including chemical warfare agents
  • Textile-like inner improves comfort
  • Typical coverall features include color coded dual zip systems, double cuffs with knitted inner cuff, and 2-piece respirator fit hood
  • Ultrasonically welded and taped seams provides a strong liquid and particle barrier
Typical areas of application include chemical handling/transportation, oil-based mud protection, hazardous waste remediation, sewage purification installations, industrial/tank cleaning, HAZMAT emergency response (i.e. Level B), pharamaceutical, mining, and agriculture.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B, EN ISO 13982-1 Type 5-B, ANSI/ISEA 101.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty