Would you like to log in again?
VWR® AC Round Bottles, Clear, Wide Mouth
Supplier: VWR
VWR®
293063 293071 293090 293066 293091 293056 293081 293061 293080 293073 VWR89043-864 VWR89043-866 293086 293058 293095 293068 293087 293077
10861-990CS 812 USD InStock
10861-990 10862-233 10862-255 10862-244 10862-256 10862-242 10862-259 10862-226 10862-358 10862-248 10862-238 10862-356 10862-247 10861-988 10862-230 10862-362 10862-360 10862-250
VWR® AC Round Bottles, Clear, Wide Mouth
Bottles Glass Bottles
VWR® Clear, wide mouth AC round bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • No graduations
  • Available in a variety of package sizes with different closure options
These bottles do not feature graduations.
They may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures in separate bags, or in bulk packs without closures.
Packaging: Bottles are available in three packaging options.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty